MilliporeSigma
  • Nrg1β Released in Remote Ischemic Preconditioning Improves Myocardial Perfusion and Decreases Ischemia/Reperfusion Injury via ErbB2-Mediated Rescue of Endothelial Nitric Oxide Synthase and Abrogation of Trx2 Autophagy.
Nrg1β Released in Remote Ischemic Preconditioning Improves Myocardial Perfusion and Decreases Ischemia/Reperfusion Injury via ErbB2-Mediated Rescue of Endothelial Nitric Oxide Synthase and Abrogation of Trx2 Autophagy.
Arteriosclerosis, thrombosis, and vascular biology (2021-05-28)
Venkatesh Kundumani-Sridharan, Jaganathan Subramani, Cade Owens, Kumuda C Das
RESUMEN
[Figure: see text].
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-fosfotirosina, clon 4G10 ®, clone 4G10®, Upstate®, from mouse
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad