Merck
27-Hydroxycholesterol regulates human SLC22A12 gene expression through estrogen receptor action.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-12-29)
Masaya Matsubayashi, Yoshihiko M Sakaguchi, Yoshiki Sahara, Hitoki Nanaura, Sotaro Kikuchi, Arvand Asghari, Linh Bui, Shinko Kobashigawa, Mari Nakanishi, Riko Nagata, Takeshi K Matsui, Genro Kashino, Masatoshi Hasegawa, Shin Takasawa, Masahiro Eriguchi, Kazuhiko Tsuruya, Shushi Nagamori, Kazuma Sugie, Takahiko Nakagawa, Minoru Takasato, Michihisa Umetani, Eiichiro Mori
RESUMEN
The excretion and reabsorption of uric acid both to and from urine are tightly regulated by uric acid transporters. Metabolic syndrome conditions, such as obesity, hypercholesterolemia, and insulin resistance, are believed to regulate the expression of uric acid transporters and decrease the excretion of uric acid. However, the mechanisms driving cholesterol impacts on uric acid transporters have been unknown. Here, we show that cholesterol metabolite 27-hydroxycholesterol (27HC) upregulates the uric acid reabsorption transporter URAT1 encoded by SLC22A12 via estrogen receptors (ER). Transcriptional motif analysis showed that the SLC22A12 gene promoter has more estrogen response elements (EREs) than other uric acid reabsorption transporters such as SLC22A11 and SLC22A13, and 27HC-activated SLC22A12 gene promoter via ER through EREs. Furthermore, 27HC increased SLC22A12 gene expression in human kidney organoids. Our results suggest that in hypercholesterolemic conditions, elevated levels of 27HC derived from cholesterol induce URAT1/SLC22A12 expression to increase uric acid reabsorption, and thereby, could increase serum uric acid levels.