Merck
  • Astragaloside IV protects against podocyte apoptosis by inhibiting oxidative stress via activating PPARγ-Klotho-FoxO1 axis in diabetic nephropathy.
Astragaloside IV protects against podocyte apoptosis by inhibiting oxidative stress via activating PPARγ-Klotho-FoxO1 axis in diabetic nephropathy.
Life sciences (2021-01-22)
Lina Xing, Ji Fang, Bingbing Zhu, Li Wang, Junliang Chen, Yunman Wang, Jiebo Huang, Hao Wang, Xingmei Yao
RESUMEN
Podocyte apoptosis plays an important role in the pathogenesis of diabetic nephropathy (DN). Astragaloside IV (AS-IV) has been shown to protect against podocyte apoptosis. Here we aim to investigate the mechanism responsible for the protective effects of AS-IV. Diabetic db/db mice and high glucose (HG)-cultured podocytes were treated with AS-IV. Renal function and histopathological changes were measured to evaluate the therapeutic effects of AS-IV against DN. Adenovirus-mediated Klotho overexpression, Klotho siRNA, and PPARγ inhibitor were applied in vitro to investigate the potential mechanism. The expression levels of mRNA and proteins were analyzed by qRT-PCR, western blot or immunofluorescence. Intracellular ROS and mitochondrial superoxide were detected by DHE and MitoSOx Red, respectively. Cell apoptosis was evaluated by TUNEL staining and flow cytometry. AS-IV improved renal function and ameliorated podocyte injury in db/db mice accompanied with enhanced Klotho expression in glomerular podocytes. In vitro, AS-IV inhibited HG-induced podocyte apoptosis and restored HG-inhibited Klotho expression, whereas Klotho knockdown abrogated the anti-apoptosis action of AS-IV. Further study showed that adenovirus-mediated Klotho overexpression enhanced Forkhead transcription factor O1 (FoxO1)-dependent antioxidant activity and attenuated HG-evoked oxidative stress and apoptosis. AS-IV prevented HG-induced FoxO1 inhibition and oxidative stress, whereas Klotho knockdown reversed these effects. Cotreatment with PPARγ inhibitor T0070907 abolished AS-IV-induced Klotho expression and anti-apoptosis action. These data suggested that AS-IV attenuated podocyte apoptosis presumably by inhibiting oxidative stress via activating PPARγ-Klotho-FoxO1 signaling pathway, thereby ameliorating DN. This study provided new insights into the molecular mechanisms of AS-IV against DN.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human KL