Merck
Effects of conformational restriction of 2-amino-3-benzoylthiophenes on A(1) adenosine receptor modulation.
Journal of medicinal chemistry (2010-09-03)
Luigi Aurelio, Celine Valant, Bernard L Flynn, Patrick M Sexton, Jonathan M White, Arthur Christopoulos, Peter J Scammells
RESUMEN
2-Amino-3-benzoylthiophenes (2A3BTs) have been widely reported to act as allosteric enhancers (AEs) at the A(1) adenosine receptor (A(1)AR). Herein we describe the synthesis of a series of 1-aminoindeno[1,2-c]thiophen-8-ones and a series of (2-aminoindeno[2,1-b]thiophen-3-yl)(phenyl)methanones as conformationally rigid analogues of the 2A3BTs. These compounds were screened using a functional assay of A(1)AR-mediated phosphorylation of extracellular signal-regulated kinases 1 and 2 (ERK1/2) in intact Chinese hamster ovary (CHO) cells to identify both potential agonistic effects as well as the ability to allosterically modulate the activity of the orthosteric agonist, N(6)-(R-phenylisopropyl)adenosine (R-PIA). All of the 1-aminoindeno[1,2-c]thiophen-8-ones (14a-c and 17a-f) proved either to be inactive or behaved as antagonists in the functional assay. However, the (2-aminoindeno[2,1-b]thiophen-3-yl)(phenyl)methanones with para-chloro substitution (compounds 25b, 25d, and 25f) did significantly augment the R-PIA response, indicating a positive allosteric effect.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Aminobenzophenone, 98%