Merck
PTPS Facilitates Compartmentalized LTBP1 S-Nitrosylation and Promotes Tumor Growth under Hypoxia.
Molecular cell (2019-10-20)
Qin Zhao, Ke Zheng, Chunmin Ma, Jingjie Li, Lingang Zhuo, Wenhua Huang, Tao Chen, Yuhui Jiang
RESUMEN
GTP cyclohydrolase I (GTPCH), 6-pyruvoyltetrahydropterin synthase (PTPS), and sepiapterin reductase (SR) are sequentially responsible for de novo synthesis of tetrahydrobiopterin (BH4), a known co-factor for nitric oxide synthase (NOS). The implication of BH4-biosynthesis process in tumorigenesis remains to be investigated. Here, we show that PTPS, which is highly expressed in early-stage colorectal cancer, is phosphorylated at Thr 58 by AMPK under hypoxia; this phosphorylation promotes PTPS binding to LTBP1 and subsequently drives iNOS-mediated LTBP1 S-nitrosylation through proximal-coupling BH4 production within the PTPS/iNOS/LTBP1 complex. In turn, LTBP1 S-nitrosylation results in proteasome-dependent LTBP1 protein degradation, revealing an inverse relationship between PTPS pT58 and LTBP1 stability. Physiologically, the repressive effect of PTPS on LTBP1 leads to impaired transforming growth factor β (TGF-β) secretion and thereby maintains tumor cell growth under hypoxia. Our findings illustrate a molecular mechanism underlying the regulation of LTBP1-TGF-β signaling by the BH4-biosynthesis pathway and highlight the specific requirement of PTPS for tumor growth.