Merck
  • Growth Arrest-Specific 6 (GAS6) Promotes Prostate Cancer Survival by G1 Arrest/S Phase Delay and Inhibition of Apoptosis During Chemotherapy in Bone Marrow.
Growth Arrest-Specific 6 (GAS6) Promotes Prostate Cancer Survival by G1 Arrest/S Phase Delay and Inhibition of Apoptosis During Chemotherapy in Bone Marrow.
Journal of cellular biochemistry (2016-05-07)
Eunsohl Lee, Ann M Decker, Frank C Cackowski, Lulia A Kana, Kenji Yumoto, Younghun Jung, Jingcheng Wang, Laura Buttitta, Todd M Morgan, Russell S Taichman
RESUMEN
Prostate cancer (PCa) is known to develop resistance to chemotherapy. Growth arrest-specific 6 (GAS6), plays a role in tumor progression by regulating growth in many cancers. Here, we explored how GAS6 regulates the cell cycle and apoptosis of PCa cells in response to chemotherapy. We found that GAS6 is sufficient to significantly increase the fraction of cells in G1 and the duration of phase in PCa cells. Importantly, the effect of GAS6 on G1 is potentiated during docetaxel chemotherapy. GAS6 altered the levels of several key cell cycle regulators, including the downregulation of Cyclin B1 (G2 /M phase), CDC25A, Cyclin E1, and CDK2 (S phase entry), while the upregulation of cell cycle inhibitors p27 and p21, Cyclin D1, and CDK4. Importantly, these changes became further accentuated during docetaxel treatment in the presence of GAS6. Moreover, GAS6 alters the apoptotic response of PCa cells during docetaxel chemotherapy. Docetaxel induced PCa cell apoptosis is efficiently suppressed in PCa cell culture in the presence of GAS6 or GAS6 secreted from co-cultured osteoblasts. Similarly, the GAS6-expressing bone environment protects PCa cells from apoptosis within primary tumors in vivo studies. Docetaxel induced significant levels of Caspase-3 and PARP cleavage in PCa cells, while GAS6 protected PCa cells from docetaxel-induced apoptotic signaling. Together, these data suggest that GAS6, expressed by osteoblasts in the bone marrow, plays a significant role in the regulation of PCa cell survival during chemotherapy, which will have important implications for targeting metastatic disease. J. Cell. Biochem. 117: 2815-2824, 2016. © 2016 Wiley Periodicals, Inc.