Merck
FGFR4 regulates tumor subtype differentiation in luminal breast cancer and metastatic disease.
The Journal of clinical investigation (2020-06-24)
Susana Garcia-Recio, Aatish Thennavan, Michael P East, Joel S Parker, Juan M Cejalvo, Joseph P Garay, Daniel P Hollern, Xiaping He, Kevin R Mott, Patricia Galván, Cheng Fan, Sara R Selitsky, Alisha R Coffey, David Marron, Fara Brasó-Maristany, Octavio Burgués, Joan Albanell, Federico Rojo, Ana Lluch, Eduardo Martinez de Dueñas, Jeffery M Rosen, Gary L Johnson, Lisa A Carey, Aleix Prat, Charles M Perou
RESUMEN
Mechanisms driving tumor progression from less aggressive subtypes to more aggressive states represent key targets for therapy. We identified a subset of luminal A primary breast tumors that give rise to HER2-enriched (HER2E) subtype metastases, but remain clinically HER2 negative (cHER2-). By testing the unique genetic and transcriptomic features of these cases, we developed the hypothesis that FGFR4 likely participates in this subtype switching. To evaluate this, we developed 2 FGFR4 genomic signatures using a patient-derived xenograft (PDX) model treated with an FGFR4 inhibitor, which inhibited PDX growth in vivo. Bulk tumor gene expression analysis and single-cell RNA sequencing demonstrated that the inhibition of FGFR4 signaling caused molecular switching. In the Molecular Taxonomy of Breast Cancer International Consortium (METABRIC) breast cancer cohort, FGFR4-induced and FGFR4-repressed signatures each predicted overall survival. Additionally, the FGFR4-induced signature was an independent prognostic factor beyond subtype and stage. Supervised analysis of 77 primary tumors with paired metastases revealed that the FGFR4-induced signature was significantly higher in luminal/ER+ tumor metastases compared with their primaries. Finally, multivariate analysis demonstrated that the FGFR4-induced signature also predicted site-specific metastasis for lung, liver, and brain, but not for bone or lymph nodes. These data identify a link between FGFR4-regulated genes and metastasis, suggesting treatment options for FGFR4-positive patients, whose high expression is not caused by mutation or amplification.