Merck
Analgesic and antioxidant activity of mangiferin and its derivatives: the structure activity relationship.
Biological & pharmaceutical bulletin (2005-04-02)
Ahsana Dar, Shaheen Faizi, Sabira Naqvi, Talat Roome, Sadia Zikr-ur-Rehman, Muhammad Ali, Sadiqa Firdous, Syed Tarique Moin
RESUMEN
Mangiferin, 2-beta-D-glucopyranosyl-1,3,6,7-tetrahydroxy-9H-xanthen-9-one, obtained directly from methanolic extracts of Bombax ceiba leaves in substantial amounts demonstrated strong antioxidant activity (EC(50) 5.8+/-0.96 mug/ml or 13.74 muM) using DPPH assay comparable to rutin, commonly used as antioxidant for medical purposes. The acetyl and cinnamoyl derivatives were found to be less active than mangiferin whereas, methyl and 3,6,7-trimethylether tetraacetate derivatives were inactive implying that for antioxidant activity, free hydroxyl groups and catechol moiety are essential. Moreover, mangiferin showed hepatoprotective activity against carbon tetrachloride induced liver injury further supporting the free radical scavenging property in the in vivo system. Additionally, plant extracts and mangiferin failed to exhibit acute anti-inflammatory activity whereas, it displayed significant analgesic effect in acetic acid-induced writhing and hot plate tests in mice. Using naloxone, it was revealed that plant extracts induced analgesia was independent of opioid receptor, whereas, mangiferin demonstrated significant interaction with it at peripheral site with a slight contribution at the neuronal level.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Mangiferin Mangifera indica