Merck
  • L-(+)-2-Amino-4-thiophosphonobutyric acid (L-thioAP4), a new potent agonist of group III metabotropic glutamate receptors: increased distal acidity affords enhanced potency.
L-(+)-2-Amino-4-thiophosphonobutyric acid (L-thioAP4), a new potent agonist of group III metabotropic glutamate receptors: increased distal acidity affords enhanced potency.
Journal of medicinal chemistry (2007-08-29)
Chelliah Selvam, Cyril Goudet, Nadia Oueslati, Jean-Philippe Pin, Francine C Acher
RESUMEN
L-2-Amino-4-phosphonobutyric acid (l-AP4), l-2-amino-4-thiophosphonobutyric acid (l-thioAP4), and l-2-amino-4-(hydroxy)phosphinylbutyric acid (desmethylphosphinothricin, DMPT) were synthesized from protected vinylglycine. They were tested as agonists at group III metabotropic glutamate receptors (mGluR) along with phosphinothricin (PT). DMPT and PT display a much lower potency at mGlu4 receptor (EC50 = 4.0 and 1100 microM, respectively) in comparison to l-AP4 (EC50 = 0.08 microM), whereas l-thioAP4 has a 2-fold higher potency (EC50 = 0.039 microM). Similar rank orders of potency were observed at mGlu6,7 and mGlu8 receptors. The higher potency of l-thioAP4 is due to its stronger second acidity compared to l-AP4. These pKa values of 5.56 and 6.88, respectively, were determined using 31P NMR chemical shift variations. The second distal negative charge of l-AP4/l-thioAP4 probably provides stronger binding to specific basic residues of the binding sites of group III mGluRs, which stabilizes the active conformation of the receptor.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-(+)-2-Amino-4-phosphonobutyric acid, optical purity optical purity: ≥95% (HPLC, Marfey′s reagent)