Merck
Effects of antidepressants in rats trained to discriminate the beta-2 adrenergic agonist clenbuterol.
Pharmacology, biochemistry, and behavior (1999-06-17)
M M Makhay, J M O'Donnell
RESUMEN
The ability of indirectly acting agonists such as norepinephrine uptake inhibitors, serotonin reuptake inhibitors, and atypical antidepressants to substitute for clenbuterol, a beta-2 adrenergic agonist, was examined in rats trained to discriminate 0.03 mg/kg clenbuterol and saline using a fixed-ratio 10 (FR 10) schedule with water reinforcement. The beta-2 selective adrenergic agonist clenbuterol produced an orderly dose-response relationship, and its discriminative stimulus effects were antagonized by the beta-adrenergic antagonist propranolol. It was found that the effects of tricyclic antidepressants and selective norepinephrine uptake inhibitors did not generalize to the discriminative stimulus effects of clenbuterol, with the exception of high doses of protriptyline. Moreover, compounds from other drug classes, including fluoxetine and phenelzine, did not substitute for clenbuterol. Atypical antidepressants, such as trazodone, rolipram, and bupropion also did not engender drug-appropriate responding. Prenalterol and dobutamine, both purported to be beta-1 adrenergic receptor agonists, partially substituted for clenbuterol, but at relatively high doses. The present results show that the antidepressants tested do not share discriminative stimulus effects with clenbuterol, a beta-2 adrenergic agonist.