Merck
  • mTORC1 coordinates an immediate unfolded protein response-related transcriptome in activated B cells preceding antibody secretion.
mTORC1 coordinates an immediate unfolded protein response-related transcriptome in activated B cells preceding antibody secretion.
Nature communications (2020-02-07)
Brian T Gaudette, Derek D Jones, Alexandra Bortnick, Yair Argon, David Allman
RESUMEN
How activated B cells build biosynthetic pathways and organelle structures necessary for subsequent robust antibody secretion is still unclear. The dominant model holds that nascent plasma cells adapt to increased antibody synthesis by activating the unfolded protein response (UPR) under the control of the transcription factor Xbp1. Here, by analyzing gene expression in activated B cells with or without plasma cell-inductive signals, we find that follicular B cells up-regulate a wide array of UPR-affiliated genes before initiating antibody secretion; furthermore, initial transcription of these loci requires the mTORC1 kinase adaptor, Raptor, but not Xbp1. Transcriptomic analyses of resting marginal zone B cells, which generate plasma cells with exceptionally rapid kinetics, reinforce these results by revealing the basal expression of UPR-affiliated mRNA networks without detectable Xbp1 activity. We thus conclude that B cells utilize mTORC1 to prepare for subsequent plasma cell function, before the onset of antibody synthesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Millipore
Placa de 96 pocillos Multiscreen®, membrana de PVDF hidrófoba, pore size 0.45 μm, non-sterile