Merck
Untargeted lipidomics using liquid chromatography-ion mobility-mass spectrometry reveals novel triacylglycerides in human milk.
Scientific reports (2020-06-11)
Alexandra D George, Melvin C L Gay, Mary E Wlodek, Robert D Trengove, Kevin Murray, Donna T Geddes
RESUMEN
Human milk provides the infant with the essential nutritive and non-nutritive factors required for health, growth and development. The human milk lipidome is complex, but comprises predominantly triacylglycerides. Historically, the fatty acid profile of the entire human milk lipidome has been investigated, and many relationships have been identified between infant health and fatty acids. Most of these fatty acids are, however, delivered to the infant as triacylglycerides. Using liquid chromatography-ion mobility-mass spectrometry, the objective of this study was to characterise the triacylglyceride profile of human milk and elucidate relationships between the triacylglyceride profile and infant outcomes in a cohort of 10 exclusively breastfeeding woman-infant dyads. 205 triacylglycerides were identified, including 98 previously not reported in human milk. The dose of specific triacylglycerides differed in relation to infant health, such as lauric acid containing TAGs, which were delivered in significantly higher dose to healthy infants compared to unwell infants.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tripentadecanoin, ≥99%