Merck
  • A highly efficient palladium/copper cocatalytic system for direct arylation of heteroarenes: an unexpected effect of Cu(Xantphos)I.
A highly efficient palladium/copper cocatalytic system for direct arylation of heteroarenes: an unexpected effect of Cu(Xantphos)I.
Journal of the American Chemical Society (2010-02-26)
Jinkun Huang, Johann Chan, Ying Chen, Christopher J Borths, Kyle D Baucom, Robert D Larsen, Margaret M Faul
RESUMEN
Heteroarenes are important structural moieties in many chemical industry fields. A highly efficient Pd/Cu-catalyzed C-H arylation method for a range of heterocycles has been discovered. It was found that the key to the success of this transformation is a combination of a palladium catalyst and a well-defined copper cocatalyst. The efficiency and low loadings of catalyst (0.25 mol %) and cocatalyst (1 mol %) together with the mild reaction conditions demonstrate this method to be practically useful and mechanistically interesting.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Iodo[4,5-bis(diphenylphosphino)-9,9-dimethylxanthene]copper(I)