Merck
PEG-8 Laurate Fermentation of Staphylococcus epidermidis Reduces the Required Dose of Clindamycin Against Cutibacterium acnes.
International journal of molecular sciences (2020-07-28)
Shinta Marito, Sunita Keshari, Chun-Ming Huang
RESUMEN
The probiotic activity of skin Staphylococcus epidermidis (S. epidermidis) bacteria can elicit diverse biological functions via the fermentation of various carbon sources. Here, we found that polyethylene glycol (PEG)-8 Laurate, a carbon-rich molecule, can selectively induce the fermentation of S. epidermidis, not Cutibacterium acnes (C. acnes), a bacterium associated with acne vulgaris. The PEG-8 Laurate fermentation of S. epidermidis remarkably diminished the growth of C. acnes and the C. acnes-induced production of pro-inflammatory macrophage-inflammatory protein 2 (MIP-2) cytokines in mice. Fermentation media enhanced the anti-C. acnes activity of a low dose (0.1%) clindamycin, a prescription antibiotic commonly used to treat acne vulgaris, in terms of the suppression of C. acnes colonization and MIP-2 production. Furthermore, PEG-8 Laurate fermentation of S. epidermidis boosted the activity of 0.1% clindamycin to reduce the sizes of C. acnes colonies. Our results demonstrated, for the first time, that the PEG-8 Laurate fermentation of S. epidermidis displayed the adjuvant effect on promoting the efficacy of low-dose clindamycin against C. acnes. Targeting C. acnes by lowering the required doses of antibiotics may avoid the risk of creating drug-resistant C. acnes and maintain the bacterial homeostasis in the skin microbiome, leading to a novel modality for the antibiotic treatment of acne vulgaris.