Merck
  • Toward fingerprinting cellular lipidomes directly from biological samples by two-dimensional electrospray ionization mass spectrometry.
Toward fingerprinting cellular lipidomes directly from biological samples by two-dimensional electrospray ionization mass spectrometry.
Analytical biochemistry (2004-06-19)
Xianlin Han, Jingyue Yang, Hua Cheng, Hongping Ye, Richard W Gross
RESUMEN
Cellular lipids are composed of linear combinations of aliphatic chains covalently attached to backbones and/or head groups that collectively represent the cellular lipidome. Herein we present a novel two-dimensional electrospray ionization mass spectrometric approach to fingerprint most of the major and many of the minor lipid classes in the hepatic cellular lipidome, which collectively represent >80% of the total lipid mass, directly from their chloroform extracts. Through lipid class-selective intrasource ionization and subsequent analysis of two-dimensional cross-peak intensities, the chemical identity and mass composition of individual molecular species of most mouse hepatic lipid classes were determined from its chloroform extract. This new integrated platform provides a robust foundation for the automated analysis of multidimensional mass spectra to advance the level of analytical scripting in lipidomics to a new plateau.
MATERIALES
Número de producto
Marca
Descripción del producto
Avanti
15:0 PE, 1,2-dipentadecanoyl-sn-glycero-3-phosphoethanolamine, powder