MilliporeSigma
Gas-chromatographic study on the stereoselectivity of deprenyl metabolism.
Journal of pharmaceutical and biomedical analysis (1998-07-31)
Z Tarjányi, H Kalász, G Szebeni, I Hollósi, M Báthori, S Fürst
RESUMEN
The metabolism and urinary elimination of both (-)-deprenyl and (+)-deprenyl have been studied. Gas-chromatographic analysis with mass specific detection indicated that the metabolism of (-)-deprenyl results in a large excess of methamphetamine compared to amphetamine, while the metabolism of (+)-deprenyl gave nearly equal amounts of amphetamine and methamphetamine. A novel deprenyl metabolite, phenylacetone, was also identified in our studies.