Merck
Determination of beer aroma compounds using headspace solid-phase microcolumn extraction.
Talanta (2010-11-03)
Ján Hrivňák, Daniela Smogrovičová, Pavol Nádaský, Jana Lakatošová
RESUMEN
A rapid sampling technique for the analysis of beer aroma compounds is described. The headspace (10 ml) is passed through the microcolumn filled with 5mg of Tenax TA and thermally desorbed in a modified GC inlet (modification is described). Eight compounds (from acetaldehyde to 2-phenylethanol) in four beer samples were analyzed. The correlation coefficients (r(2)), repeatability (RSD) and limits of detection (LOD) were 0.9973-0.9994, 2.1-6.9% and 0.00002-0.13 mg/l, respectively. The methodology can be useful for routine beer sample analysis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Ethyl hexanoate, analytical standard