Merck
A Simple Method for Ion Channel Recordings Using Fine Gold Electrode.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2016-12-13)
Daichi Okuno, Minako Hirano, Hiroaki Yokota, Yukiko Onishi, Junya Ichinose, Toru Ide
RESUMEN
The artificial bilayer single-channel recording technique is commonly used to observe detailed pharmacological properties of various ion channel proteins. It permits easy control of the solution and membrane lipid composition, and is also compatible with pharmacological screening devices. However, its use is limited due to low measurement efficiency. Here, we develop a novel artificial bilayer single-channel recording technique in which bilayers are made and channels are reconstituted into the membranes by contacting a gold electrode to the lipid-solution interface. Using this technique, we measured the single-channel currents of two channel-forming peptides, gramicidin and alamethicin, and a channel-forming protein, α-hemolysin. This technique requires only one action, allowing the technique to potentially be combined with high-throughput screening devices.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Thiol-dPEG®4-acid, >95% (HPLC)