Merck
  • Nucleotides released from Aβ₁₋₄₂ -treated microglial cells increase cell migration and Aβ₁₋₄₂ uptake through P2Y₂ receptor activation.
Nucleotides released from Aβ₁₋₄₂ -treated microglial cells increase cell migration and Aβ₁₋₄₂ uptake through P2Y₂ receptor activation.
Journal of neurochemistry (2012-02-23)
Hye Jung Kim, Deepa Ajit, Troy S Peterson, Yanfang Wang, Jean M Camden, W Gibson Wood, Grace Y Sun, Laurie Erb, Michael Petris, Gary A Weisman
RESUMEN
Amyloid β-protein (Aβ) deposits in brains of Alzheimer's disease patients generate proinflammatory cytokines and chemokines that recruit microglial cells to phagocytose Aβ. Nucleotides released from apoptotic cells activate P2Y(2) receptors (P2Y(2) Rs) in macrophages to promote clearance of dead cells. In this study, we investigated the role of P2Y(2) Rs in the phagocytosis and clearance of Aβ. Treatment of mouse primary microglial cells with fibrillar (fAβ(1-42) ) and oligomeric (oAβ(1-42) ) Aβ(1-42) aggregation solutions caused a rapid release of ATP (maximum after 10 min). Furthermore, fAβ(1-42) and oAβ(1-42) treatment for 24 h caused an increase in P2Y(2) R gene expression. Treatment with fAβ(1-42) and oAβ(1-42) aggregation solutions increased the motility of neighboring microglial cells, a response inhibited by pre-treatment with apyrase, an enzyme that hydrolyzes nucleotides. The P2Y(2) R agonists ATP and UTP caused significant uptake of Aβ(1-42) by microglial cells within 30 min, which reached a maximum within 1 h, but did not increase Aβ(1-42) uptake by primary microglial cells isolated from P2Y(2) R(-/-) mice. Inhibitors of α(v) integrins, Src and Rac decreased UTP-induced Aβ(1-42) uptake, suggesting that these previously identified components of the P2Y(2) R signaling pathway play a role in Aβ phagocytosis by microglial cells. Finally, we found that UTP treatment enhances Aβ(1-42) degradation by microglial cells, but not in cells isolated from P2Y(2) R(-/-) mice. Taken together, our findings suggest that P2Y(2) Rs can activate microglial cells to enhance Aβ clearance and highlight the P2Y(2) R as a therapeutic target in Alzheimer's disease.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
[Gln22]-Amyloid β 1-40 human, ≥95% (HPLC)