Merck
Quantitative and Dynamic Catalogs of Proteins Released during Apoptotic and Necroptotic Cell Death.
Cell reports (2020-01-30)
Maria C Tanzer, Annika Frauenstein, Che A Stafford, Kshiti Phulphagar, Matthias Mann, Felix Meissner
RESUMEN
The inflammatory functions of the cytokine tumor necrosis factor (TNF) rely on its ability to induce cytokine production and to induce cell death. Caspase-dependent and caspase-independent pathways-apoptosis and necroptosis, respectively-regulate immunogenicity by the release of distinct sets of cellular proteins. To obtain an unbiased, systems-level understanding of this important process, we here applied mass spectrometry-based proteomics to dissect protein release during apoptosis and necroptosis. We report hundreds of proteins released from human myeloid cells in time course experiments. Both cell death types induce receptor shedding, but only apoptotic cells released nucleosome components. Conversely, necroptotic cells release lysosomal components by activating lysosomal exocytosis at early stages of necroptosis-induced membrane permeabilization and show reduced release of conventionally secreted cytokines.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Millipore
Membrana DE PVDF Immobilon®- P, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anticuerpo anti-MLKL, clon 3H1, clone 3H1, from rat
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Yoduro de propidio, ≥94% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-Cathepsin B (Ab-1) Mouse mAb (CA10), lyophilized, clone CA10, Calbiochem®
En este momento no podemos mostrarle ni los precios ni la disponibilidad