Merck
Aromatic versus Aliphatic α-Diimine Ligands in Heteroleptic Copper(I) Emitters: Photophysical and Electrochemical Properties.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2019-11-07)
Masaki Yoshida, Sae Yanagida, Daisuke Saito, Atsushi Kobayashi, Masako Kato
RESUMEN
The electrochemical and photophysical properties of a heteroleptic Cu(I) complex bearing an aliphatic α-diimine ligand, [Cu(dab)(xantphos)]+ (Cu-dab; dab = N,N'-diphenyl-2,3-dimethyl-1,4-diazabutadiene, xantphos = 4,5-bis(diphenylphosphino)-9,9-dimethylxanthene), were evaluated together with those of complexes [Cu(dmp)(xantphos)]+ (Cu-dmp; dmp = 2,9-dimethyl-1,10-phenanthroline), [Cu(dmbpy)(xantphos)]+ (Cu-dmbpy; dmbpy = 5,5'-dimethyl-2,2'-bipyridine), and [Cu(bq)(xantphos)]+ (Cu-bq; bq = 2,2'-biquinoline), bearing aromatic diimine ligands. Cu-dab exhibited a two-step ligand-centered redox behavior, where the first wave corresponded to an electrochemically reversible one-electron reduction process. Although Cu(I)-aromatic diimine complexes Cu-dmp, Cu-dmbpy, and Cu-bq exhibited obvious luminescence from the metal-to-ligand charge transfer (MLCT) excited state, Cu-dab did not show any luminescence. Computational studies indicated that this non-luminescent property was caused by the large structural relaxation of Cu-dab during photoexcitation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Xantphos, 97%
Sigma-Aldrich
5,5′-Dimethyl-2,2′-dipyridyl, 98%