Merck
  • Knockdown of PLCB2 expression reduces melanoma cell viability and promotes melanoma cell apoptosis by altering Ras/Raf/MAPK signals.
Knockdown of PLCB2 expression reduces melanoma cell viability and promotes melanoma cell apoptosis by altering Ras/Raf/MAPK signals.
Molecular medicine reports (2019-11-21)
Huahui Zhang, Tao Xie, Yongjie Shui, Yiying Qi
RESUMEN
Malignant melanoma has the highest malignancy rate among all skin cancer and is characterized by an insidious onset, high invasion and poor patient prognosis. Yet, the mechanisms involved remain unclear and warrant further investigation. Based on bioinformatic analysis, phospholipase C β2 (PLCB2) has been found to be correlated with melanoma growth. The present study was the first to demonstrate that PLCB2 is a key factor affecting melanoma proliferation and apoptosis. Here, microarray datasets from the publicly available Gene Expression Omnibus (GEO) database were employed, and gene set enrichment analysis (GSEA) was introduced to identify candidate transcription factors. PLCB2 was identified as a crucial gene in the protein‑protein interaction (PPI) network. The expression of PLCB2 mRNA in various cancer lines was analyzed by reverse transcription‑polymerase chain reaction (RT‑PCR). In addition, the proliferation ability and apoptosis rate in human melanoma cells overexpressing or not overexpressing PLCB2 were assessed using colony formation assay, flow cytometry and the Cell Counting Kit‑8 (CCK‑8) assay. Cell viability and apoptosis‑related factors, such as p53, Bcl‑2, Bax and caspase‑3 were significantly regulated. Knockdown of PLCB2 suppressed the activation of the Ras/Raf/MAPK signaling pathway. In conclusion, knockdown of PLCB2 suppressed cell viability and promoted cell apoptosis by activating the Ras/Raf/MAPK pathway. Thus, PLCB2 may utilized as a potential therapeutic target in patients with melanoma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human PLCB2