Merck
  • MSTN Attenuates Cardiac Hypertrophy through Inhibition of Excessive Cardiac Autophagy by Blocking AMPK /mTOR and miR-128/PPARγ/NF-κB.
MSTN Attenuates Cardiac Hypertrophy through Inhibition of Excessive Cardiac Autophagy by Blocking AMPK /mTOR and miR-128/PPARγ/NF-κB.
Molecular therapy. Nucleic acids (2020-01-11)
Hanping Qi, Jing Ren, Lina Ba, Chao Song, Qianhui Zhang, Yonggang Cao, Pilong Shi, Bowen Fu, Yongsheng Liu, Hongli Sun
RESUMEN
Cardiac hypertrophy, a response of the heart to increased workload, is a major risk factor for heart failure. Myostatin (MSTN) is an inhibitor of myogenesis, regulating the number and size of skeletal myocytes. In recent years, cardiomyocyte autophagy also has been considered to be involved in controlling the hypertrophic response. However, less is known about the detailed mechanism of MSTN on cardiac hypertrophy via regulation of cardiomyocyte autophagy. In this study, we found that the deletion of MSTN potentiated abdominal aorta coarctation (AAC) and angiotensin II (Ang II)-induced pathological cardiac hypertrophy and cardiac autophagy; however, AAC and Ang II-induced cardiac hypertrophic phenotype and cardiac autophagy were dramatically diminished by MSTN in vivo and in vitro. Mechanistically, the anti-hypertrophic and anti-autophagic effects mediated by MSTN in response to pathological stimuli were associated with the direct inactivation of activated protein kinase (AMPK)/mammalian target of rapamycin (mTOR) and activation of the peroxisome proliferator-activated receptor gamma (PPARγ)/nuclear factor κB (NF-κB) signaling pathway. Additionally, miR-128 aggravated the progression of cardiac hypertrophy through suppressing its target PPARγ. Furthermore, MSTN downregulated miR-128 expression induced by AAC and Ang II. Taken together, MSTN significantly blunts pathological cardiac hypertrophy and dysfunction, at least in part, by inhibiting excessive cardiac autophagy via blocking AMPK/mTOR and miR-128/PPARγ/NF-κB signaling pathways.