Merck
Members of the EpCAM signalling pathway are expressed in gastric cancer tissue and are correlated with patient prognosis.
British journal of cancer (2013-09-07)
V S Warneke, H-M Behrens, J Haag, S Krüger, E Simon, M Mathiak, M P A Ebert, C Röcken
RESUMEN
We investigated the expression of members of the epithelial cell adhesion molecule (EpCAM) signalling pathway in gastric cancer (GC) testing the following hypotheses: are these molecules expressed in GC and are they putatively involved in GC biology. The study cohort consisted of 482 patients. The following members of the EpCAM signalling pathway were analysed by immunohistochemistry and were correlated with various clinico-pathological patient characteristics: extracellular domain of EpCAM (EpEX), intracellular domain of EpCAM (EpICD), E-cadherin, β-catenin, presenilin-2 (PSEN2), and ADAM17. All members of the EpCAM signalling pathway were differentially expressed in GC. The expression correlated significantly with tumour type (EpEX, EpICD, E-cadherin, β-catenin, and PSEN2), mucin phenotype (EpEX, EpICD, β-catenin, and ADAM17), T-category (EpEX, E-cadherin, and β-catenin), N-category (EpEX and β-catenin), UICC tumour stage (EpEX, EpICD, β-catenin, and PSEN2), tumour grade (EpEX, EpICD, E-cadherin, β-catenin, and PSEN2), and patients' survival (EpEX, EpICD, and PSEN2). A significant coincidental expression in GC was found for EpEX, EpICD, E-cadherin, β-catenin, PSEN2, and ADAM17. Decreased immunodetection of EpEX in locally advanced GC was not associated with decreased EpCAM mRNA levels. All members of the EpCAM signalling pathway are expressed in GC. The expression correlated significantly with each other and with various clinico-pathological patient characteristics, including patients' survival. Thus, the EpCAM signalling pathway is a highly interesting putative therapeutic target in GC.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-MSH2 Antibody, clone FE11, clone FE11, from mouse