Merck
BDNF locally potentiates GABAergic presynaptic machineries: target-selective circuit inhibition.
Cerebral cortex (New York, N.Y. : 1991) (2004-07-09)
Shizu Ohba, Takamitsu Ikeda, Yuji Ikegaya, Nobuyoshi Nishiyama, Norio Matsuki, Maki K Yamada
RESUMEN
Inhibitory neurotransmission is critical for neuronal circuit formation. To examine whether inhibitory neurotransmission receives target-selective modulation in the long term, we expressed the cDNA of brain-derived neurotrophic factor (BDNF), which has been shown to induce the augmentation of GABAergic synapses in vivo and in vitro, in a small population of cultured hippocampal neurons. At 48 h after transfection, the expression level of glutamic acid decarboxylase 65 (GAD65), a GABA synthetic enzyme that resides mainly in GABAergic terminals, was selectively enhanced around the BDNF-expressing neurons, in comparison with the neighboring control neurons interposed between the BDNF-expressing neurons and inhibitory neurons. Exogenous BDNF application for 48 h also increased the GAD level and enhanced the GABA release probability. These potentiating effects were attenuated in inhibitory synapses on neurons expressing a dominant negative form of the BDNF receptor (tTrkB). This suggests that postsynaptic BDNF-TrkB signaling contributes to the target-selective potentiation of inhibitory presynaptic machineries. Since BDNF is expressed in an activity-dependent manner in vivo, this selectivity may be one of the key mechanisms by which the independence of functional neuronal circuits is maintained.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-GABA A Receptor β2/3 Antibody, clone 62-3G1, Upstate®, from mouse