Merck
Transcription factor AP-2α promotes EGF-dependent invasion of human trophoblast.
Endocrinology (2011-02-10)
Katarzyna Biadasiewicz, Stefan Sonderegger, Peter Haslinger, Sandra Haider, Leila Saleh, Christian Fiala, Jürgen Pollheimer, Martin Knöfler
RESUMEN
The basic helix-span-helix transcription factor activating protein (AP)-2α is critically involved in cell-specific hormone expression of syncytializing human trophoblasts. Its role in invasive trophoblast differentiation, however, remains largely elusive. Using RT-PCR, Western blotting, and immunofluorescence of first-trimester placentae, we here show that AP-2α is expressed in extravillous trophoblasts (EVTs) both in situ and in vitro as well as in invasive trophoblast cell lines. Its protein expression was increased upon supplementation of epidermal growth factor (EGF) both in primary EVTs and trophoblastic SGHPL-5 cells. Gene silencing of AP-2α using small hairpin microRNA (shRNAmir) did not affect basal invasion of SGHPL-5 cells through Matrigel-coated filters but reduced EGF-stimulated invasion. Similarly, treatment of primary EVTs with AP-2α small interfering RNA decreased EGF-dependent invasion. Proliferation of SGHPL-5 cells and primary EVTs, measured by cumulative cell numbers and 5-bromo-2'-deoxyuridine labeling, respectively, were not affected on loss of AP-2α. EGF-dependent induction of matrix metalloproteinase (MMP)-2, pro- and active form of urokinase plasminogen activator, and chorionic gonadotropin (CG)-β was noticed in shRNAmir-control cells, whereas these genes were suppressed in EGF-treated shRNAmir-AP-2α cells. Similarly, EGF-stimulated MMP-2 and CGβ protein expression was reduced in AP-2α small interfering RNA-treated primary EVTs. Knockdown of AP-2α also decreased luciferase activity of the CGβ5 promoter in SGHPL-5 cells, which was compensated upon transient overexpression of AP-2α cDNA. In conclusion, we show that AP-2α expression positively affects human trophoblast invasion under EGF-stimulated conditions, likely by inducing critical invasion-promoting genes such MMP-2, urokinase plasminogen activator, and CG.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse