Merck
Colistin-oxolinic acid blood agar: a selective medium for the isolation of Gardnerella vaginalis.
Journal of clinical microbiology (1985-05-01)
J S Thompson
RESUMEN
Colistin-oxolinic acid medium is proposed as a selective isolation medium for Gardnerella vaginalis. The medium is effective in inhibiting staphylococci and gram-negative bacteria while allowing growth of G. vaginalis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Oxolinic acid, quinolone antibiotic