Merck
Synthesis and screening of a library of Re/Tc-based amyloid probes derived from beta-breaker peptides.
Bioconjugate chemistry (2008-04-15)
Karin A Stephenson, Leslie C Reid, Jon Zubieta, John W Babich, Mei-Ping Kung, Hank F Kung, John F Valliant
RESUMEN
Through the development and application of a unique approach for producing Re-metallopeptides, a new class of peptide-derived probes that are designed to target beta-amyloid plaques was developed. Derivatives of a class of beta-breaker peptides having the core sequence lvffa or affvl (lower case letters represent D-amino acids) and the single amino acid chelate quinoline (SAACQ) ligand which can bind Re and (99m)Tc were prepared on an automated peptide synthesizer. Both monomeric and dimeric peptides were synthesized in modest to good yields where in select examples a biotin-containing amino acid derivative was included to act as a linker point for further conjugation to carrier proteins. The Re complexes for all reported peptides were prepared similarly and screened for their ability to inhibit fibrillogenesis. Two of the reported compounds showed excellent inhibitory properties (8a: 40 +/- 5% amyloid formation versus control; 16: 40 +/- 4%) and warrant further investigation. For one of these leads, the (99m)Tc analogue was synthesized and the product showed high stability toward histidine and cysteine challenges, making it a viable candidate for in vivo biodistribution studies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fmoc-D-Lys(Biotin)-OH, ≥95%