Merck
Maternal vitamin C regulates reprogramming of DNA methylation and germline development.
Nature (2019-09-06)
Stephanie P DiTroia, Michelle Percharde, Marie-Justine Guerquin, Estelle Wall, Evelyne Collignon, Kevin T Ebata, Kathryn Mesh, Swetha Mahesula, Michalis Agathocleous, Diana J Laird, Gabriel Livera, Miguel Ramalho-Santos
RESUMEN
Development is often assumed to be hardwired in the genome, but several lines of evidence indicate that it is susceptible to environmental modulation with potential long-term consequences, including in mammals1,2. The embryonic germline is of particular interest because of the potential for intergenerational epigenetic effects. The mammalian germline undergoes extensive DNA demethylation3-7 that occurs in large part by passive dilution of methylation over successive cell divisions, accompanied by active DNA demethylation by TET enzymes3,8-10. TET activity has been shown to be modulated by nutrients and metabolites, such as vitamin C11-15. Here we show that maternal vitamin C is required for proper DNA demethylation and the development of female fetal germ cells in a mouse model. Maternal vitamin C deficiency does not affect overall embryonic development but leads to reduced numbers of germ cells, delayed meiosis and reduced fecundity in adult offspring. The transcriptome of germ cells from vitamin-C-deficient embryos is remarkably similar to that of embryos carrying a null mutation in Tet1. Vitamin C deficiency leads to an aberrant DNA methylation profile that includes incomplete demethylation of key regulators of meiosis and transposable elements. These findings reveal that deficiency in vitamin C during gestation partially recapitulates loss of TET1, and provide a potential intergenerational mechanism for adjusting fecundity to environmental conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido L-ascórbico, 99%