Merck
Extraction and high-performance liquid chromatographic method for the determination of microcystins in raw and treated waters.
The Analyst (1994-07-01)
L A Lawton, C Edwards, G A Codd
RESUMEN
Increasing concern over the presence of microcystins (cyanobacterial/blue-green algal hepatotoxins) in water supplies has emphasized the need for a suitable analytical method. As many microcystins are known to exist, a method was developed that permits the determination of numerous variants by a single procedure. The method involves filtration to separate cyanobacterial cells from water, allowing intracellular and extracellular toxin levels to be assessed. The cellular components of the samples are extracted repeatedly in methanol, which was found to be the most versatile solvent tested for the extraction of microcystins. The efficiency of this extraction procedure was found to be independent of cell biomass. The filtered water was subjected to trace enrichment using a C18 solid-phase extraction cartridge, followed by identification and determination by photodiode-array high-performance liquid chromatography. The procedure was assessed using four water samples (two raw and two treated) spiked with a mixture of five microcystins and the cyanobacterial hepatotoxin nodularin. Recoveries of all but one microcystin were found to be good when spiked with concentrations as low as 250 ng l-1. The linearity and precision of the experimental procedure were assessed for five microcystins and nodularin. The proposed method permits rapid sample processing and determination of several microcystins.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Microcystin-LW, Microcystis aeruginosa, Analog of Microcystin-LR with Trp substituted in place of Arg. Believed to be more cell-permeable than other microcystins making it useful for biochemical studies in intact cells.