- Generation of an integration-free induced pluripotent stem cell line, FJMUi001-A, from a hereditary spastic paraplegia patient carrying compound heterozygous p.P498L and p.R618W mutations in CAPN1 (SPG76).
Generation of an integration-free induced pluripotent stem cell line, FJMUi001-A, from a hereditary spastic paraplegia patient carrying compound heterozygous p.P498L and p.R618W mutations in CAPN1 (SPG76).
Stem cell research (2019-01-06)
Ying-Qian Lu, En-Lin Dong, Wei-Qi Yang, Lu-Lu Lai, Xiao-Hong Lin, Li-Xiang Ma, Wan-Jin Chen, Ning Wang, Xiang Lin
PMID30611022
RESUMEN
The human iPS cell line, hiPS-SPG76 (FJMUi001-A), derived from skin fibroblasts from a 42-year-old male hereditary spastic paraplegia patient carrying compound heterozygous p.P498L (c.1493C > T) and p.R618W (c.1852C > T) mutations in the CAPN1 gene, was generated by non-integrative reprogramming vectors encoding OCT3/4, SOX2, KLF4, and c-MYC. The established hiPS-SPG76 was free of genomically integrated reprogramming genes, had a normal karyotype, expressed pluripotency markers, and had capacity to form three germ layers in vitro and in vivo. This generated hiPS cell line offers a useful resource to study the pathogenesis of SPG76.