Merck
Copper(II) complexes as potential anticancer and Nonsteroidal anti-inflammatory agents: In vitro and in vivo studies.
Scientific reports (2019-03-29)
Afzal Hussain, Mohamed Fahad AlAjmi, Md Tabish Rehman, Samira Amir, Fohad Mabood Husain, Ali Alsalme, Maqsood Ahmad Siddiqui, Abdulaziz A AlKhedhairy, Rais Ahmad Khan
RESUMEN
Copper-based compounds are promising entities for target-specific next-generation anticancer and NSAIDS therapeutics. In lieu of this, benzimidazole scaffold plays an important role, because of their wide variety of potential functionalizations and coordination modes. Herein, we report three copper complexes 1-3 with benzimidazole-derived scaffolds, a biocompatible molecule, and secondary ligands viz, 1-10-phenanthroline and 2,2'-bipyridyl. All the copper complexes have been designed, synthesized and adequately characterized using various spectroscopic techniques. In-vitro, human serum albumin (HSA) binding was also carried out using fluorescence technique and in-silico molecular modeling studies, which exhibited significant binding affinities of the complexes with HSA. Furthermore, copper complexes 1-3 were tested for biological studies, i.e., anticancer as well as NSAIDS. In vitro cytotoxicity results were carried out on cultured MCF-7 cell lines. To get the insight over the mechanism of action, GSH depletion and change in lipid peroxidation were tested and thus confirmed the role of ROS generation, responsible for the cytotoxicity of the complexes 1-3. Moreover, the copper complexes 1-3 were tested for potential to act as NSAIDS on albino rats and mice in animal studies in-vivo. Additionally, we also predicted the mechanism of action of the copper complexes 1-3 using molecular modeling studies with COX-2 inhibitor.