- Two Clostridiumperfringens Type E Isolates in France.
Two Clostridiumperfringens Type E Isolates in France.
Toxins (2019-03-03)
Laure Diancourt, Jean Sautereau, Alexis Criscuolo, Michel R Popoff
PMID30823654
RESUMEN
Clostridiumperfringens type E is a less frequently isolated C.perfringens type and has not previously been reported in France. We have characterized two recent type E isolates, C.perfringens 508.17 from the intestinal content of a calf that died of enterotoxemia, and 515.17 from the stool of a 60-year-old woman, subsequent to food poisoning, which contained the plasmid pCPPB-1 with variant iota toxin and C. perfringens enterotoxin genes.