Merck
Antimicrobial biopolymer formation from sodium alginate and algae extract using aminoglycosides.
PloS one (2019-03-27)
Lokender Kumar, John Brice, Linda Toberer, Judith Klein-Seetharaman, Daniel Knauss, Susanta K Sarkar
RESUMEN
Antimicrobial biopolymers provide a biodegradable, sustainable, safe, and cheap approach to drug delivery and wound dressing to control bacterial infection and improve wound healing respectively. Here, we report a one-step method of making antimicrobial alginate polymer from sodium alginate and aqueous extract of Wakame using antibiotic aminoglycosides. Thin layer chromatography of commercially available sodium alginate and Wakame extract showed similar oligosaccharide profiles. Screening of six aminoglycosides showed that kanamycin disulfate and neomycin sulfate produces the highest amount of biopolymer; however, kanamycin disulfate produces the most malleable and form fitting biopolymer. Image texture analysis of biopolymers showed similar quantification parameters for all the six aminoglycosides. Weight of alginate polymer as a function of aminoglycoside concentration follows a growth model of prion protein, consistent with the aggregating nature of both processes. Slow release of antibiotics and the resulting zone of inhibition against E. coli DH5α were observed by agar well diffusion assay. Inexpensive method of production and slow release of antibiotics will enable diverse applications of antimicrobial alginate biopolymer reported in this paper.