Merck
β-Catenin-RAS interaction serves as a molecular switch for RAS degradation via GSK3β.
EMBO reports (2018-11-11)
Sang-Kyu Lee, Woo-Jeong Jeong, Yong-Hee Cho, Pu-Hyeon Cha, Jeong-Su Yoon, Eun Ji Ro, Sooho Choi, Jeong-Min Oh, Yunseok Heo, Hyuntae Kim, Do Sik Min, Gyoonhee Han, Weontae Lee, Kang-Yell Choi
RESUMEN
RAS proteins play critical roles in various cellular processes, including growth and transformation. RAS proteins are subjected to protein stability regulation via the Wnt/β-catenin pathway, and glycogen synthase kinase 3 beta (GSK3β) is a key player for the phosphorylation-dependent RAS degradation through proteasomes. GSK3β-mediated RAS degradation does not occur in cells that express a nondegradable mutant (MT) β-catenin. Here, we show that β-catenin directly interacts with RAS at the α-interface region that contains the GSK3β phosphorylation sites, threonine 144 and threonine 148 residues. Exposure of these sites by prior β-catenin degradation is required for RAS degradation. The introduction of a peptide that blocks the β-catenin-RAS interaction by binding to β-catenin rescues the GSK3β-mediated RAS degradation in colorectal cancer (CRC) cells that express MT β-catenin. The coregulation of β-catenin and RAS stabilities by the modulation of their interaction provides a mechanism for Wnt/β-catenin and RAS-ERK pathway cross-talk and the synergistic transformation of CRC by both APC and KRAS mutations.