Merck
Antifungal activity and Shore A hardness of a tissue conditioner incorporated with terpinen-4-ol and cinnamaldehyde.
Clinical oral investigations (2019-05-22)
Laura de Fátima Souto Maior, Panmella Pereira Maciel, Victor Yuri Nicolau Ferreira, Cíntia de Lima Gouveia Dantas, Jeferson Muniz de Lima, Lúcio Roberto Cançado Castellano, André Ulisses Dantas Batista, Paulo Rogério Ferreti Bonan
RESUMEN
This study investigated the anti-Candida activity and the Shore A hardness of a tissue conditioner (Softone™) modified by incorporation of terpinen-4-ol and cinnamaldehyde. Agar diffusion, microdilution, and mechanism of action methods were performed to determine to evaluate the antifungal activity of phytoconstituents. Then, phytoconstituents in varying concentrations were incorporated into the tissue conditioner. The anti-Candida effect of the modified conditioner was evaluated through agar punch well and biofilm formation methods. Shore A hardness of the experimental liners was evaluated after baseline, 24 h, 48 h, 4 days, and 7 days immersion on artificial saliva. The phytoconstituents incorporated into Softone showed completely inhibited fungal growth in concentrations of 20-40% and did not present significant antifungal activity until their concentrations where higher than 5%. There were differences between non-modified Softone and M5, M10, C10, and T10% (p < 0.05). The groups containing 10-40% of cinnamaldehyde incorporated into Softone were able to completely inhibit the biofilm. Concentrations below 40% of terpinen-4-ol showed unsatisfactory biofilm inhibition. The T40% and C40% groups presented the lowest Shore A hardness values. Hardness values from groups T40% at 7 days (p = 0.476); C40% at 4 days (p = 0.058); and T20% (p = 0.058), C20% (p = 0.205), T30% (p = 0.154), and C30% (p = 0.874) after 48 h did not differ from the control group. Cinnamaldehyde incorporated into Softone inhibited Candida biofilm formation at concentrations of 10-40%, being more effective than terpinen-4-ol modification despite of halo inhibition observed by both products. All modifications showed a very similar pattern of hardness being useful for clinical practice.