Merck
  • The Low Toxicity of Graphene Quantum Dots is Reflected by Marginal Gene Expression Changes of Primary Human Hematopoietic Stem Cells.
The Low Toxicity of Graphene Quantum Dots is Reflected by Marginal Gene Expression Changes of Primary Human Hematopoietic Stem Cells.
Scientific reports (2019-08-21)
Stefan Fasbender, Lisa Zimmermann, Ron-Patrick Cadeddu, Martina Luysberg, Bastian Moll, Christoph Janiak, Thomas Heinzel, Rainer Haas
RESUMEN
Graphene quantum dots (GQDs) are a promising next generation nanomaterial with manifold biomedical applications. For real world applications, comprehensive studies on their influence on the functionality of primary human cells are mandatory. Here, we report the effects of GQDs on the transcriptome of CD34+ hematopoietic stem cells after an incubation time of 36 hours. Of the 20 800 recorded gene expressions, only one, namely the selenoprotein W, 1, is changed by the GQDs in direct comparison to CD34+ hematopoietic stem cells cultivated without GQDs. Only a meta analysis reveals that the expression of 1171 genes is weakly affected, taking into account the more prominent changes just by the cell culture. Eight corresponding, weakly affected signaling pathways are identified, which include, but are not limited to, the triggering of apoptosis. These results suggest that GQDs with sizes in the range of a few nanometers hardly influence the CD34+ cells on the transcriptome level after 36 h of incubation, thereby demonstrating their high usability for in vivo studies, such as fluorescence labeling or delivery protocols, without strong effects on the functional status of the cells.