Merck
Characterization of an OrtT-like toxin of Salmonella enterica serovar Houten.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology] (2019-05-06)
Luiz Carlos Bertucci Barbosa, Rodolfo Dos Santos Carrijo, Milena Barbosa da Conceição, Jonatas Erick Maimoni Campanella, Edson Crusca Júnior, Thais Oliveira Secches, Maria Celia Bertolini, Reinaldo Marchetto
RESUMEN
The Escherichia coli GhoT/GhoS system is a type V toxin-antitoxin system in which the antitoxin GhoS cleaves the GhoT mRNA, controlling its translation. GhoT is a small hydrophobic protein that damages bacterial membranes. OrtT is a GhoT-like toxin, but it apparently lacks a corresponding antitoxin and serves a different physiologic role. Using a profile hidden Markov model approach, a Salmonella enterica serovar Houten genome was screened to obtain homologs of GhoT/OrtT. We only found one protein (referred to here as OrtT-Sal) that shared more sequence identity with OrtT than GhoT. The chromosomal region around the coding sequence of OrtT-Sal suggests that it is an orphan toxin and can be under RpoH activation. To study OrtT-Sal, we chemically synthesized and expressed in E. coli the whole toxin and its N- and C-terminal regions (OrtT-Sal1-29 and OrtT-Sal29-57, respectively). Our findings have shown that the overproduction of the polypeptides resulted in severe growth inhibition and cell lysis. Using circular dichroism, we found that OrtT-Sal, OrtT-Sal1-29, and OrtT-Sal29-57 form an alpha-helical structure in the presence of SDS micelles or TFE. Finally, using carboxyfluorescein-loaded lipid vesicles, we determined that the polypeptides damage lipid membrane directly.