Merck
  • In vitro inhibitory effects of major bioactive constituents of Andrographis paniculata, Curcuma longa and Silybum marianum on human liver microsomal morphine glucuronidation: A prediction of potential herb-drug interactions arising from andrographolide, curcumin and silybin inhibition in humans.
In vitro inhibitory effects of major bioactive constituents of Andrographis paniculata, Curcuma longa and Silybum marianum on human liver microsomal morphine glucuronidation: A prediction of potential herb-drug interactions arising from andrographolide, curcumin and silybin inhibition in humans.
Drug metabolism and pharmacokinetics (2017-12-16)
Verawan Uchaipichat
RESUMEN
This study aimed to investigate the liver microsomal inhibitory effects of silybin, silychristin, andrographolide, and curcumin by using morphine as an in vitro UGT2B7 probe substrate, and predict the magnitude of the herb-drug interaction arising from these herbal constituents' inhibition in vivo. Studies were performed in the incubation with and without bovine serum albumin (BSA). Andrographolide and curcumin showed a marked inhibition on morphine 3- and 6-glucuronidation with IC50 of 50&87 and 96&111 μM, respectively. In the presence of 2%BSA, andrographolide also showed a strong inhibition on morphine 3- and 6-glucuronidation (IC50 4.4&21.6 μM) whereas curcumin showed moderate inhibition (IC50 338&333 μM). In the absence and presence of 2%BSA, morphine 3- and 6-glucuronidation was moderately inhibited by silybin (IC50 583&862 and 1252&1421 μM, respectively), however was weakly inhibited by silychristin (IC50 3527&3504 and 1124&1530 μM, respectively). The Ki of andrographolide, curcumin and silybin on morphine 3- and 6-glucuronidation were 7.1&9.5, 72.7&65.2, and 224.5&159.7 μM, respectively, while the respective values generated from the system containing 2%BSA were 2.4&3.1, 96.4&108.8, and 366.3&394.5 μM. Using the in vitro and in vivo extrapolation approach, andrographolide was herbal component that may have had a potential interaction in vivo when it was co-administered with morphine.