Merck
  • Silencing of PPARαBb mRNA in brown trout primary hepatocytes: effects on molecular and morphological targets under the influence of an estrogen and a PPARα agonist.
Silencing of PPARαBb mRNA in brown trout primary hepatocytes: effects on molecular and morphological targets under the influence of an estrogen and a PPARα agonist.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2018-12-12)
Tânia Vieira Madureira, Ivone Pinheiro, Fernanda Malhão, L Filipe C Castro, Eduardo Rocha, Ralph Urbatzka
RESUMEN
The crosstalk between peroxisome proliferator-activated receptor α (PPARα) and estrogenic pathways are shared from fish to humans. Salmonid fish had an additional genome duplication, and two PPARα isoforms (PPARαBa and PPARαBb) were previously identified. Since a negative regulation between estrogen signaling and PPARα was described, a post-transcriptional gene silencing for PPARαBb was designed in primary brown trout hepatocytes. The aims of the study were to: (i) decipher the effects of PPARαBb knock-down on peroxisome morphology and on mRNA expression of potential target genes, and (ii) to assess the cross-interferences caused by an estrogenic compound (17α-ethinylestradiol - EE2) and a PPARα agonist (Wy-14,643 - Wy) using the established knock-down model. A knock-down efficiency of 70% was achieved for PPARαBb and its silencing significantly reduced the volume density of peroxisomes, but did not alter mRNA levels of the studied genes. Exposure to Wy did not change peroxisome morphology or mRNA expression, but under silencing conditions Wy rescued the volume density of peroxisomes to control levels, and increased acyl-coenzyme A oxidase 1-3l (Acox1-3l) mRNA. Exposure to EE2 caused a reduction of peroxisome volume density, but under silencing conditions this effect was abolished and ApoA1 mRNA level was diminished. The morphological alterations of peroxisomes by WY and EE2 demonstrated that obtained results are PPARαBb dependent, and suggest the regulation of unknown downstream targets of PPARαBb. In summary, PPARαBb is involved in the control of peroxisome size and/or number, which opens future opportunities to explore its regulation and molecular targets.