Merck
5xFAD Mice Display Sex-Dependent Inflammatory Gene Induction During the Prodromal Stage of Alzheimer's Disease.
Journal of Alzheimer's disease : JAD (2019-07-20)
Zahra Manji, Asheebo Rojas, Wenyi Wang, Raymond Dingledine, Nicholas H Varvel, Thota Ganesh
RESUMEN
Alzheimer's disease (AD) pathology consists of extracellular deposits of amyloid-β peptides (Aβ) and intracellular neurofibrillary tangles. These pathological alterations are accompanied by a neuroinflammatory response consisting of increased expression of inflammatory mediators. An anti-inflammatory strategy designed to prevent or delay the development of AD would benefit from knowing when neuroinflammation appears in the transgenic models during prodromal disease stages relative to Aβ pathology. We investigated the expression patterns of inflammatory mediators in the brain of 5xFAD mice in comparison to development of Aβ deposition. Expression changes in inflammatory mediators and glial markers are more robust in female mice starting at three months of age, in contrast to males in which there is no clear trend through five months. Female and male 5xFAD mice also displayed an age-dependent increase in cortical Aβ deposition congruent with neuroinflammation. Thus, in the 5xFAD mouse model of AD, administration of an anti-inflammatory agent would be most efficacious when administered before three months of age.