Merck
  • Chronic administration of the histamine H3 receptor agonist immepip decreases L-Dopa-induced dyskinesias in 6-hydroxydopamine-lesioned rats.
Chronic administration of the histamine H3 receptor agonist immepip decreases L-Dopa-induced dyskinesias in 6-hydroxydopamine-lesioned rats.
Psychopharmacology (2019-02-15)
Alberto Avila-Luna, Camilo Ríos, Arturo Gálvez-Rosas, Sergio Montes, José-Antonio Arias-Montaño, Antonio Bueno-Nava
RESUMEN
Histamine H3 receptors (H3Rs) are co-expressed with dopamine D1 receptors (D1Rs) by striato-nigral medium spiny GABAergic neurons, where they functionally antagonize D1R-mediated responses. We examined whether the chronic administration of the H3R agonist immepip modifies dyskinesias induced by L-3,4-dihydroxyphenylalanine, L-Dopa (LIDs), in rats lesioned with 6-hydroxydopamine in the substantia nigra pars compacta, and the effect of D1R and H3R co-activation on glutamate and GABA content in dialysates from the dorsal striatum of naïve rats. The systemic administration (i.p.) of L-Dopa for 14 days significantly increased axial, limb, and orolingual abnormal involuntary movements (AIMs) compared with the vehicle group. The chronic administration of the H3R agonist immepip alongside L-Dopa significantly decreased axial, limb, and orolingual AIMs compared with L-Dopa alone, but AIMs returned to previous values on immepip withdrawal. Chronic immepip was ineffective when administered prior to L-Dopa. The chronic administration of immepip significantly decreased GABA and glutamate content in striatal dialysates, whereas the administration of L-Dopa alone increased GABA and glutamate content. These results indicate that chronic H3R activation reduces LIDs, and the effects on striatal GABA and glutamate release provide evidence for a functional interaction between D1Rs and H3Rs.