Merck
Investigation of Polyurethane Matrix Membranes for Salivary Nitrate ISFETs to Prevent the Drift.
Sensors (Basel, Switzerland) (2019-06-20)
Shuto Osaki, Takuya Kintoki, Takayo Moriuchi-Kawakami, Kenichi Kitamura, Shin-Ichi Wakida
RESUMEN
We have investigated human-stress monitoring by making use of salivary nitrate, which can be a candidate for stress markers, with ion-selective field-effect transistors (ISFETs). ISFETs are suitable for on-site single-drop analysis of salivary nitrate within 10 s. However, when ISFETs are used for salivary nitrate, ISFETs have a problem that is called the initial drift. The initial drift makes accurate nitrate monitoring difficult. Thus, the purpose of this study is to prevent the initial drift and to search for a new, simple polymer to possess a better performance of sensor responses than conventional matrix membranes, such as PVC. In this research, we investigated ISFETs using specific matrix membranes, for example KP-13, Pellethane®--, and P7281-PU. The initial drift was evaluated from the fluctuations of the response values generated by the ISFETs when immersed in saliva or aqueous solution. As a result, P7281-PU showed a prevention effect on the initial drift, both in the whole saliva and in various solutions. Furthermore, the cause of drift may be H+ diffusion, and the drift prevention effect of P7281-PU may be affected by urethane bond capturing H+ in the ion-selective membrane. This result suggests that a continuous nitrate monitoring is feasible and may be applied to wearable sensors.