Merck
In vitro culture of mammalian inner ear hair cells.
Journal of Zhejiang University. Science. B (2018-09-07)
Lu-Wen Zhang, Xiao-Hui Cang, Ye Chen, Min-Xin Guan
RESUMEN
Auditory function in vertebrates depends on the transduction of sound vibrations into electrical signals by inner ear hair cells. In general, hearing loss resulting from hair cell damage is irreversible because the human ear has been considered to be incapable of regenerating or repairing these sensory elements following severe injury. Therefore, regeneration and protection of inner ear hair cells have become an exciting, rapidly evolving field of research during the last decade. However, mammalian auditory hair cells are few in number, experimentally inaccessible, and barely proliferate postnatally in vitro. Various in vitro primary culture systems of inner ear hair cells have been established by different groups, although many challenges remain unresolved. Here, we briefly explain the structure of the inner ear, summarize the published methods of in vitro hair cell cultures, and propose a feasible protocol for culturing these cells, which gave satisfactory results in our study. A better understanding of in vitro hair cell cultures will substantially facilitate research involving auditory functions, drug development, and the isolation of critical molecules involved in hair cell biology.