Merck
Influence of Matrigel on Single- and Multiple-Spheroid Cultures in Breast Cancer Research.
SLAS discovery : advancing life sciences R & D (2019-03-22)
Madalina Andreea Badea, Mihaela Balas, Anca Hermenean, Alina Ciceu, Hildegard Herman, Daniela Ionita, Anca Dinischiotu
RESUMEN
This study aimed to develop and compare single and multiple 3D models such as multicellular tumor spheroids and to investigate the influence of Matrigel on their morphological and functional behavior. MDA-MB-231 3D models were generated in the presence and absence of Matrigel and their key biological properties within 6 days of culture were monitored. Our results revealed the formation of well-defined 3D models in the presence of Matrigel, with a uniform morphology, increased diameter, good circularity, and increased expression of a proliferation marker (PCNA). In comparison, 3D models generated without Matrigel were characterized by an irregular border, reduced dimensions and circularity, and a decrease of PCNA expression. Similarities between the single and multiple 3D cultures were found in their viability, Nrf2 expression, and glutathione (GSH) content. The influence of Matrigel on MDA-MB-231 spheroids metabolism under hypoxic conditions was highlighted by released lactate dehydrogenase and nitric oxide, GSH levels and expression of Nrf2 and Hsp70 proteins. Based on the increased expression of PCNA and the development of the hypoxia process in the presence of extracellular matrix, our study showed that the addition of Matrigel improves the growing environment of tumor spheroids, making it closer to that of in vivo tumor conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-β-actina, anticuerpo monoclonal, clone AC-15, purified from hybridoma cell culture