Merck
Low expression of miR-186-5p regulates cell apoptosis by targeting toll-like receptor 3 in high glucose-induced cardiomyocytes.
Journal of cellular biochemistry (2018-12-07)
Ye Liu, Wei Zheng, Yue Pan, Jing Hu
RESUMEN
To investigate the effect and mechanism of microRNA-186-5p (miR-186-5p) on the apoptosis in high glucose (HG)-treated cardiomyocytes. Diabetic cardiomyopathy model was established in cardiomyocytes by stimulating with HG. The expressions of miR-186-5p and toll-like receptor 3 (TLR3) were detected by quantitative polymerase chain reaction or Western blot analysis, respectively. Apoptosis was detected in HG-treated cardiomyocytes by flow cytometry and Western blot analysis. The interaction between miR-186-5p and TLR3 was explored by bioinformatics analysis and luciferase activity assay. Results showed that miR-186-5p expression was downregulated in HG-treated cardiomyocytes and its overexpression reversed HG-induced apoptosis and cleaved caspase-3 protein expression. Moreover, TLR3 was indicated as a target of miR-186-5p and regulated by miR-186-5p. Knockdown of TLR3 suppressed HG-induced apoptosis and cleaved caspase-3 protein expression. Besides, restoration of TLR3 ablated the effect of miR-186-5p on cell apoptosis. Collectively, miR-186-5p attenuated HG-induced apoptosis by regulating TLR3 in cardiomyocytes, providing novel biomarker for treatment of diabetic cardiomyopathy.