Merck
Chrysophanic acid blocks proliferation of colon cancer cells by inhibiting EGFR/mTOR pathway.
Phytotherapy research : PTR (2010-11-23)
Myung Sun Lee, Eun Young Cha, Ji Young Sul, In Sang Song, Ji Yeon Kim
RESUMEN
Inactivation of epidermal growth factor receptor (EGFR) is a prime method used in colon cancer therapy. Here it is shown that chrysophanic acid, a natural anthraquinone, has anticancer activity in EGFR-overexpressing SNU-C5 human colon cancer cells. Chrysophanic acid preferentially blocked proliferation in SNU-C5 cells but not in other cell lines (HT7, HT29, KM12C, SW480, HCT116 and SNU-C4) with low levels of EGFR expression. Chrysophanic acid treatment in SNU-C5 cells inhibited EGF-induced phosphorylation of EGFR and suppressed activation of downstream signaling molecules, such as AKT, extracellular signal-regulated kinase (ERK) and the mammalian target of rapamycin (mTOR)/ribosomal protein S6 kinase (p70S6K). Chrysophanic acid (80 and 120 µm) significantly blocked cell proliferation when combined with the mTOR inhibitor, rapamycin. These findings offer the first evidence of anticancer activity for chrysophanic acid via EGFR/mTOR mediated signaling transduction pathway.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,8-Dihydroxy-3-methylanthraquinone, 98%
Supelco
Chrysophanol, analytical standard