Merck
  • A rapid method for the preparation of 125I-labelled human growth hormone for receptor studies, using reverse-phase high performance liquid chromatography.
A rapid method for the preparation of 125I-labelled human growth hormone for receptor studies, using reverse-phase high performance liquid chromatography.
Biochemical and biophysical research communications (1986-01-29)
M M Ilondo, I Dehart, P De Meyts
RESUMEN
Human growth hormone was labelled with 125 Iodine by the stoichiometric modification of the chloramine-T method to a specific activity of 50-80 microCi/microgram, and the iodinated mixture was purified by reverse-phase high performance liquid chromatography using a C18 column (SynChropak RP-P) and a linear gradient. Compared with the usual Sephadex G-100 chromatography, HPLC gave a much better separation, with a higher yield and a considerably reduced analysis time (30 min vs 5 h). The [125I]-labelled preparation had normal binding to IM-9 lymphocyte receptors. The maximum bindability of the HPLC-purified preparation approximated 90%, which is the best value so far reported for human growth hormone. It is concluded that HPLC is a fast, convenient and reproducible method for obtaining an improved [125I]-labelled human growth hormone for receptor studies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Kit de análisis MAP cinasa/Erk, no radiactivo, MAP Kinase/Erk Assay Kit is designed to measure the phosphotransferase activity of MAP (mitogen-activated protein) Kinase in immunoprecipitates & column fractions.