Merck
Increased adult hippocampal brain-derived neurotrophic factor and normal levels of neurogenesis in maternal separation rats.
Journal of neuroscience research (2005-02-04)
Mia H Greisen, C Anthony Altar, Tom G Bolwig, Richard Whitehead, Gitta Wörtwein
RESUMEN
Repeated maternal separation of rat pups during the early postnatal period may affect brain-derived neurotrophic factor (BDNF) or neurons in brain areas that are compromised by chronic stress. In the present study, a highly significant increase in hippocampal BDNF protein concentration was found in adult rats that as neonates had been subjected to 180 min of daily separation compared with handled rats separated for 15 min daily. BDNF protein was unchanged in the frontal cortex and hypothalamus/paraventricular nucleus. Expression of BDNF mRNA in the CA1, CA3, or dentate gyrus of the hippocampus or in the paraventricular hypothalamic nucleus was not affected by maternal separation. All animals displayed similar behavioral patterns in a forced-swim paradigm, which did not affect BDNF protein concentration in the hippocampus or hypothalamus. Repeated administration of bromodeoxyuridine revealed equal numbers of surviving, newly generated granule cells in the dentate gyrus of adult rats from the 15 min or 180 min groups. The age-dependent decline in neurogenesis from 3 months to 7 months of age did not differ between the groups. Insofar as BDNF can stimulate neurogenesis and repair, we propose that the elevated hippocampal protein concentration found in maternally deprived rats might be a compensatory reaction to separation during the neonatal period, maintaining adult neurogenesis at levels equal to those of the handled rats.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Factor neurotrófico derivado del cerebro de ChemiKine, ELISA en sándwich, This ChemiKine Brain Derived Neurotrophic Factor, Sandwich ELISA is used to measure & quantify BDNF levels in Neuroscience research.