Merck
  • Wortmannin and its structural analogue demethoxyviridin inhibit stimulated phospholipase A2 activity in Swiss 3T3 cells. Wortmannin is not a specific inhibitor of phosphatidylinositol 3-kinase.
Wortmannin and its structural analogue demethoxyviridin inhibit stimulated phospholipase A2 activity in Swiss 3T3 cells. Wortmannin is not a specific inhibitor of phosphatidylinositol 3-kinase.
The Journal of biological chemistry (1995-10-27)
M J Cross, A Stewart, M N Hodgkin, D J Kerr, M J Wakelam
RESUMEN
Wortmannin and its structural analogue demethoxyviridin (DMV) have been reported to be specific inhibitors of phosphatidylinositol 3-kinase activity. Here we report that these compounds are not as selective as assumed and demonstrate inhibition of bombesin-stimulated phospholipase A2 activity by both wortmannin and DMV with an IC50 (2 nM) which is slightly more potent than the inhibition of insulin-stimulated phosphatidylinositol 3,4,5-trisphosphate generation in these cells (approximately 10nM). While it has not been possible to fully block in vitro phospholipase A2 activity with wortmannin, inhibition cannot be a consequence of inhibition of PI 3-kinase activity since bombesin fails to generate 3-phosphorylated lipids in the intact cell. Therefore, while wortmannin is indeed a PI 3-kinase inhibitor, it is not as specific as previously reported, and experimental conclusions based solely on its use should be treated with caution.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Wortmanina, (inhibidor de la PI3 cinasa), The Wortmannin, (PI3 Kinase inhibitor) controls the biological activity of PI3 Kinase. This small molecule/inhibitor is primarily used for Biochemicals applications.